Assay Details
| Gene Symbol: Ppp2r5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198168.3 | NP_937811.1 | 3 - 4 | 992 | 109 | |
| GenBank mRNA | AK034167.1 | - | 3 - 4 | 1011 | 109 | |
| AK155872.1 | - | 3 - 4 | 984 | 109 | ||
| AK171041.1 | - | 3 - 4 | 963 | 109 | ||
| AK171178.1 | - | 3 - 4 | 993 | 109 | ||
| AK171430.1 | - | 3 - 4 | 1000 | 109 | ||
| BC058977.1 | - | 3 - 4 | 783 | 5687054 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5b
Genomic Map