Assay Details
| Gene Symbol: Ppp2r5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198168.3 | NP_937811.1 | 4 - 5 | 1090 | 80 | |
| GenBank mRNA | AK034167.1 | - | 4 - 5 | 1109 | 80 | |
| AK155872.1 | - | 4 - 5 | 1083 | 81 | ||
| AK171041.1 | - | 4 - 5 | 1061 | 80 | ||
| AK171178.1 | - | 4 - 5 | 1091 | 80 | ||
| AK171430.1 | - | 4 - 5 | 1098 | 80 | ||
| BC058977.1 | - | 4 - 5 | 881 | 5687054 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5b
Genomic Map