Assay Details
| Gene Symbol: Ppp2r5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198168.3 | NP_937811.1 | 8 - 9 | 1489 | 89 | |
| GenBank mRNA | AK034167.1 | - | 8 - 9 | 1508 | 89 | |
| AK155872.1 | - | 8 - 9 | 1482 | 89 | ||
| AK171041.1 | - | 8 - 9 | 1460 | 89 | ||
| AK171178.1 | - | 8 - 9 | 1490 | 89 | ||
| AK171430.1 | - | 8 - 9 | 1497 | 89 | ||
| BC026670.1 | - | 4 - 5 | 345 | 89 | ||
| BC058977.1 | - | 8 - 9 | 1280 | 5687054 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r5b
Genomic Map