Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900008885.mm.1 |
NM_023912.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008885.mm.1 |
NM_023912.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008885.mm.1 |
TC1900000983.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008884.mm.1 |
NM_023912.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008884.mm.1 |
NM_023912.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008884.mm.1 |
TC1900000983.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008886.mm.1 |
NM_023912.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008886.mm.1 |
NM_023912.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008886.mm.1 |
TC1900000983.mm.1 |
MTA TC |