Assay Details
Gene Symbol: Syvn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164709.1 | NP_001158181.1 | 4 - 5 | 586 | 70 | |
NM_028769.5 | NP_083045.4 | 4 - 5 | 953 | 70 | ||
GenBank mRNA | AK004688.2 | - | 4 - 5 | 440 | 70 | |
AK122558.1 | - | 3 - 4 | 555 | 70 | ||
AK143720.1 | - | 4 - 5 | 465 | 70 | ||
BC042199.1 | - | 4 - 5 | 448 | 4953165 | 70 | |
BC046829.1 | - | 4 - 5 | 586 | 6811574 | 70 | |
BC057917.1 | - | 4 - 5 | 867 | 5136398 | 70 | |
BC080722.1 | - | 4 - 5 | 471 | 5323695 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
