Assay Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111272.1 | NP_001104742.1 | 11 - 12 | 1612 | 114 | |
| NM_025303.3 | NP_079579.2 | 12 - 13 | 1743 | 114 | ||
| GenBank mRNA | AF459099.1 | - | 12 - 13 | 1693 | 114 | |
| AF459100.1 | - | 11 - 12 | 1562 | 114 | ||
| AK031102.1 | - | 12 - 13 | 1670 | 114 | ||
| BC010300.1 | - | 9 - 10 | 1418 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stau2
Genomic Map