Assay Details
| Gene Symbol: Stau2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111272.1 | NP_001104742.1 | 3 - 4 | 360 | 82 | |
| NM_025303.3 | NP_079579.2 | 4 - 5 | 491 | 82 | ||
| GenBank mRNA | AF459099.1 | - | 4 - 5 | 441 | 82 | |
| AF459100.1 | - | 3 - 4 | 310 | 82 | ||
| AK031102.1 | - | 4 - 5 | 419 | 82 | ||
| AK171076.1 | - | 3 - 4 | 345 | 82 | ||
| BC010300.1 | - | 1 - 2 | 166 | 82 | ||
| BC025118.1 | - | 3 - 4 | 315 | 3670067 | 82 | |
| CN665159.1 | - | 3 - 4 | 320 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stau2
Genomic Map