Assay Details
| Gene Symbol: Il18r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161842.1 | NP_001155314.1 | 4 - 5 | 759 | 85 | |
| NM_001161843.1 | NP_001155315.1 | 4 - 5 | 631 | 85 | ||
| NM_008365.2 | NP_032391.1 | 3 - 4 | 684 | 85 | ||
| GenBank mRNA | AK087129.1 | - | 3 - 4 | 633 | 85 | |
| AK089237.1 | - | 4 - 5 | 760 | 85 | ||
| AK170705.1 | - | 4 - 5 | 720 | 85 | ||
| BC020296.1 | - | 3 - 4 | 446 | 85 | ||
| BC023240.1 | - | 4 - 5 | 600 | 4483261 | 85 | |
| U43673.1 | - | 3 - 4 | 684 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il18r1
Genomic Map