Assay Details
| Gene Symbol: Mettl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207522.2 | NP_997405.2 | 10 - 11 | 1690 | 145 | |
| GenBank mRNA | AK144620.1 | - | 10 - 11 | 1682 | 145 | |
| AK148535.1 | - | 11 - 12 | 1867 | 145 | ||
| BC067068.1 | - | 10 - 11 | 1664 | 30606425 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl25
Genomic Map