Assay Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024457.2 | NP_077777.1 | 3 - 4 | 294 | 125 | |
GenBank mRNA | AK143910.1 | - | 3 - 4 | 277 | 125 | |
AK153870.1 | - | 3 - 4 | 251 | 125 | ||
AK163242.1 | - | 3 - 4 | 358 | 125 | ||
AK167548.1 | - | 3 - 4 | 250 | 125 | ||
AK178034.1 | - | 3 - 4 | 167 | 125 | ||
BC033382.1 | - | 3 - 4 | 234 | 125 | ||
BC052480.1 | - | 3 - 4 | 294 | 125 | ||
BC094066.1 | - | 3 - 4 | 234 | 125 | ||
BC147390.1 | - | 3 - 4 | 166 | 9056464 | 125 | |
BC147391.1 | - | 3 - 4 | 166 | 9056441 | 125 | |
BC147700.1 | - | 2 - 3 | 152 | 9087459 | 125 | |
BC147713.1 | - | 2 - 3 | 152 | 9087472 | 125 | |
BU848028.1 | - | 299 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
