Assay Details
| Gene Symbol: Rap1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024457.2 | NP_077777.1 | 4 - 5 | 349 | 88 | |
| GenBank mRNA | AK143910.1 | - | 4 - 5 | 332 | 88 | |
| AK153870.1 | - | 4 - 5 | 306 | 88 | ||
| AK163242.1 | - | 4 - 5 | 413 | 88 | ||
| AK167548.1 | - | 4 - 5 | 305 | 88 | ||
| AK178034.1 | - | 4 - 5 | 222 | 88 | ||
| AK196093.1 | - | 4 - 5 | 222 | 88 | ||
| AK218552.1 | - | 4 - 5 | 222 | 88 | ||
| BC033382.1 | - | 4 - 5 | 289 | 88 | ||
| BC052480.1 | - | 4 - 5 | 349 | 88 | ||
| BC094066.1 | - | 4 - 5 | 289 | 88 | ||
| BC147390.1 | - | 4 - 5 | 221 | 9056464 | 88 | |
| BC147391.1 | - | 4 - 5 | 221 | 9056441 | 88 | |
| BC147700.1 | - | 3 - 4 | 207 | 9087459 | 88 | |
| BC147713.1 | - | 3 - 4 | 207 | 9087472 | 88 | |
| BU848028.1 | - | 354 | 88 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Rap1b
Genomic Map