Assay Details
Gene Symbol: Gal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010253.3 | NP_034383.1 | 4 - 5 | 371 | 74 | |
GenBank mRNA | AK133522.1 | - | 4 - 5 | 387 | 74 | |
BC044055.1 | - | 4 - 5 | 371 | 6476459 | 74 | |
Z23069.1 | - | 2 - 3 | 150 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
