Assay Details
Gene Symbol: Uprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081189.1 | NP_001074658.1 | 4 - 5 | 748 | 94 | |
GenBank mRNA | BC147845.1 | - | 4 - 5 | 606 | 9087562 | 94 |
BC147853.1 | - | 4 - 5 | 606 | 9087570 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
