Assay Details
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011276.3 | NP_035406.3 | 3 - 4 | 474 | 128 | |
GenBank mRNA | AF069992.1 | - | 3 - 4 | 429 | 128 | |
AK013207.1 | - | 3 - 4 | 446 | 128 | ||
AK029295.1 | - | 3 - 4 | 468 | 128 | ||
BC012960.1 | - | 1 - 2 | 207 | 4193003 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
