Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900008402.mm.1 |
NM_029576.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008402.mm.1 |
NM_029576.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008402.mm.1 |
TC1900000952.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008400.mm.1 |
NM_029576.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008400.mm.1 |
NM_029576.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008400.mm.1 |
TC1900000952.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008401.mm.1 |
NM_029576.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008401.mm.1 |
NM_029576.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008401.mm.1 |
TC1900000952.mm.1 |
MTA TC |