Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900023826.mm.1 |
NM_001289645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023826.mm.1 |
NM_153420.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023826.mm.1 |
TC0900002889.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023823.mm.1 |
NM_001289645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023823.mm.1 |
NM_001289646.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023823.mm.1 |
NM_001289647.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023823.mm.1 |
NM_153420.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023823.mm.1 |
TC0900002889.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023822.mm.1 |
NM_001289645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023822.mm.1 |
NM_001289646.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023822.mm.1 |
NM_001289647.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023822.mm.1 |
NM_153420.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023822.mm.1 |
TC0900002889.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023832.mm.1 |
NM_001289645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023832.mm.1 |
NM_001289646.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023832.mm.1 |
NM_001289647.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023832.mm.1 |
NM_153420.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023832.mm.1 |
TC0900002889.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900023839.mm.1 |
NM_001289645.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023839.mm.1 |
NM_001289646.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023839.mm.1 |
NM_001289647.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023839.mm.1 |
NM_153420.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900023839.mm.1 |
TC0900002889.mm.1 |
MTA TC |