Assay Details
| Gene Symbol: Esyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028731.5 | NP_083007.2 | 15 - 16 | 1700 | 96 | |
| GenBank mRNA | AK014813.1 | - | 17 - 18 | 1810 | 96 | |
| AK044360.1 | - | 15 - 16 | 1702 | 96 | ||
| AK157352.1 | - | 15 - 16 | 1669 | 96 | ||
| BC040392.1 | - | 5 - 6 | 383 | 96 | ||
| BC052440.1 | - | 5 - 6 | 391 | 96 | ||
| BC054797.1 | - | 2 - 3 | 353 | 96 | ||
| BC059230.1 | - | 5 - 6 | 391 | 96 | ||
| BC138937.1 | - | 15 - 16 | 1609 | 8861959 | 96 | |
More Information
Set Membership: |
|

Esyt2
Genomic Map