Assay Details
| Gene Symbol: Vmn2r-ps54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_004441.1 | 1 - 2 | 72 | 88 | ||
| GenBank mRNA | AK007132.1 | - | 1 - 2 | 95 | 88 | |
| AK030272.1 | - | 1 - 2 | 73 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 788 | Zfp788 |
2810426N06Rik, AI256342, AI605193 |
-16.5kb | Sense | Chr. 7 - 41633531 - 41651534 on NCBI Build GRCm38 |
| Downstream |
vomeronasal 2, receptor 58 |
Vmn2r58 | EG628422 | 150.5kb | Anti-sense | Chr. 7 - 41836881 - 41872670 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Vmn2r-ps54
Genomic Map