Assay Details
| Gene Symbol: Imp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178601.3 | NP_848716.2 | 3 - 4 | 299 | 71 | |
| GenBank mRNA | AF334609.1 | - | 2 - 3 | 203 | 71 | |
| AK004135.1 | - | 2 - 3 | 201 | 71 | ||
| AK008178.1 | - | 3 - 4 | 261 | 71 | ||
| AK010504.1 | - | 3 - 4 | 259 | 71 | ||
| AK013334.1 | - | 3 - 4 | 228 | 71 | ||
| AK018039.1 | - | 3 - 4 | 257 | 71 | ||
| AK084933.1 | - | 3 - 4 | 235 | 71 | ||
| AK151176.1 | - | 3 - 4 | 278 | 71 | ||
| AK152214.1 | - | 3 - 4 | 278 | 71 | ||
| AK166903.1 | - | 3 - 4 | 249 | 71 | ||
| BC037602.1 | - | 2 - 3 | 220 | 71 | ||
| BC069930.1 | - | 2 - 3 | 209 | 3481801 | 71 | |
| DV649342.1 | - | 3 - 4 | 299 | 71 | ||
More Information
Set Membership: |
|

Imp4
Genomic Map