Assay Details
| Gene Symbol: Imp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178601.3 | NP_848716.2 | 8 - 9 | 850 | 69 | |
| GenBank mRNA | AF334609.1 | - | 7 - 8 | 754 | 69 | |
| AK004135.1 | - | 7 - 8 | 752 | 69 | ||
| AK008178.1 | - | 8 - 9 | 812 | 69 | ||
| AK010504.1 | - | 8 - 9 | 811 | 70 | ||
| AK013334.1 | - | 8 - 9 | 779 | 69 | ||
| AK018039.1 | - | 8 - 9 | 807 | 69 | ||
| AK084933.1 | - | 8 - 9 | 786 | 69 | ||
| AK151176.1 | - | 8 - 9 | 829 | 69 | ||
| AK152214.1 | - | 8 - 9 | 829 | 69 | ||
| AK166903.1 | - | 8 - 9 | 800 | 69 | ||
| BC037602.1 | - | 7 - 8 | 771 | 69 | ||
| BC069930.1 | - | 7 - 8 | 760 | 3481801 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20138852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20138850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20138851 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959006 | BC037602.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959006 | BC069930.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959006 | AK084933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959006 | AK151176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959006 | AK152214.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 442877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830633 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125058 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959006 | BC037602.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959006 | BC069930.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959006 | AK084933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959006 | AK151176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959006 | AK152214.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322465 | AK084933.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322465 | AK013334.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322465 | NM_178601.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20138881 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125058 | - | - |
| Whole Mouse Genome G4122A | A_51_P322465 | AK084933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322465 | AK013334.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P322465 | NM_178601.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120829 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125058 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00083513 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00556213 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00556201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00556214 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20138851 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20138852 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20138850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20138902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01063277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00778272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00209996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00556213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00778269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00083513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20138869 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10345361 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345365 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345363 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345366 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345359 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345362 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345364 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345360 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10345358 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426214_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426213_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10345357 | - | - |
| Mouse Genome 430 2.0 Array | 1426213_at | - | - |
| Mouse Genome 430 2.0 Array | 1426213_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426214_at | - | - |
| Mouse Genome 430 2.0 Array | 1426214_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27993_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27993_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100001433.mm.1 | NM_178601.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001433.mm.1 | NM_178601.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001433.mm.1 | TC0100000222.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001431.mm.1 | NM_178601.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001431.mm.1 | NM_178601.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001431.mm.1 | TC0100000222.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100001432.mm.1 | NM_178601.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001432.mm.1 | NM_178601.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100001432.mm.1 | TC0100000222.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426213_PM_at | - | - |
| Mouse Expression Set 430 | 1426214_PM_at | - | - |
| Mouse Expression Set 430 | 1426213_at | - | - |
| Mouse Expression Set 430 | 1426214_at | - | - |
| Murine Genome U74v2 Set | 95428_at | - | - |
| Murine Genome U74v2 Set | 162197_PM_at | - | - |
| Murine Genome U74v2 Set | 95428_PM_at | - | - |
| Murine Genome U74v2 Set | 162197_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426214_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426214_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426213_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426213_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426214_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426213_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426214_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426213_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3140600 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6550133 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6550133 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3140600 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003140600 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006550133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227519 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724643 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227519 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724643 | - | - |
| mouse-6 v1.1 (Illumina) | scl027993.9_8-S | - | - |
| mouse-6 v1.1 (Illumina) | scl18092.6_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6550133 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3140600 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl027993.9_8-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18092.6_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3140600 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6550133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1227519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2724643 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227519 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2724643 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl027993.9_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18092.6_1-S | - | - |

Imp4
Genomic Map