Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2379510 |
AK081638.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
AK006979.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
AK007439.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
AK010992.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
AK012569.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
AK013212.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
AK138073.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
BC049937.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
BC058725.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
BC061012.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925123 |
NM_026029.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
132734 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825606 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
613253 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
111956 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299217 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
129420 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
745102 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
655085 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
68163 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07157641 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P980532 |
AK081638.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131991 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07157641 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363459 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363455 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363457 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363463 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363448 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363456 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363465 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363467 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363461 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363466 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363453 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P08772407 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27363447 |
- |
- |