Assay Details
Gene Symbol: D230030E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045947.1 | 2 - 3 | 420 | 70 | ||
GenBank mRNA | AK084355.1 | - | 2 - 3 | 422 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | trans-acting transcription factor 4 | Sp4 |
5730497N03Rik, HF-1b, HF1-b |
-226.8kb | Sense | Chr. 12 - 118231686 - 118301440 on NCBI Build GRCm38 |
Downstream | trans-acting transcription factor 8 | Sp8 |
D930049B17Rik, mBtd |
316.2kb | Anti-sense | Chr. 12 - 118845840 - 118852578 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
