Assay Details
| Gene Symbol: Ppp1ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031868.2 | NP_114074.1 | 4 - 5 | 566 | 163 | |
| GenBank mRNA | AK007932.1 | - | 4 - 5 | 567 | 163 | |
| AK028392.1 | - | 4 - 5 | 556 | 163 | ||
| AK090070.1 | - | 4 - 5 | 584 | 163 | ||
| AK150344.1 | - | 4 - 5 | 584 | 163 | ||
| AK151582.1 | - | 4 - 5 | 584 | 163 | ||
| AK152051.1 | - | 4 - 5 | 584 | 163 | ||
| AK152585.1 | - | 4 - 5 | 584 | 163 | ||
| AK152667.1 | - | 4 - 5 | 562 | 163 | ||
| AK153517.1 | - | 4 - 5 | 548 | 163 | ||
| AK158681.1 | - | 4 - 5 | 522 | 163 | ||
| AK159575.1 | - | 4 - 5 | 574 | 163 | ||
| AK167244.1 | - | 4 - 5 | 567 | 163 | ||
| AK167538.1 | - | 4 - 5 | 579 | 163 | ||
| AK167880.1 | - | 4 - 5 | 568 | 163 | ||
| AK167981.1 | - | 4 - 5 | 567 | 163 | ||
| BC014828.1 | - | 4 - 5 | 572 | 4239005 | 163 | |
| U25809.1 | - | 4 - 5 | 587 | 163 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1ca
Genomic Map