Assay Details
Gene Symbol: Lhcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013582.2 | NP_038610.1 | 7 - 8 | 619 | 87 | |
GenBank mRNA | AK030325.1 | - | 7 - 8 | 634 | 87 | |
AK054331.1 | - | 6 - 7 | 456 | 87 | ||
M81310.1 | - | 7 - 8 | 619 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
