Assay Details
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023630.2 | NP_076119.2 | 3 - 4 | 297 | 68 | |
GenBank mRNA | AF250835.1 | - | 3 - 4 | 265 | 68 | |
AK005881.1 | - | 3 - 4 | 272 | 68 | ||
BC050756.1 | - | 3 - 4 | 280 | 6741860 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
