| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900007513.mm.1 |
NR_045304.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007513.mm.1 |
NR_045304.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007513.mm.1 |
TC1900000875.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900007518.mm.1 |
NR_045304.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007518.mm.1 |
NR_045304.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007518.mm.1 |
TC1900000875.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900007514.mm.1 |
NR_045304.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007514.mm.1 |
NR_045304.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007514.mm.1 |
TC1900000875.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900007517.mm.1 |
NR_045304.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007517.mm.1 |
NR_045304.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900007517.mm.1 |
TC1900000875.mm.1 |
MTA TC |