Assay Details
| Gene Symbol: 4930486I03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045734.1 | 1 - 2 | 247 | 86 | ||
| GenBank mRNA | AK015632.1 | - | 1 - 2 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | polycystic kidney and hepatic disease 1 | Pkhd1 |
AI118496, AI182499, FPC, Tigm1 |
Anti-sense | Chr. 1 - 20057779 - 20618082 on NCBI Build GRCm38 | |
| Downstream | microRNA 206 | Mir206 |
Mirn206, mmu-mir-206 |
297.8kb | Sense | Chr. 1 - 20679010 - 20679082 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930486I03Rik
Genomic Map