Assay Details
Gene Symbol: Gm28154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034407.1 | - | 3 - 4 | 3995 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 28784 | Gm28784 | -442.0kb | Anti-sense | Chr. 1 - 17168402 - 17173109 on NCBI Build GRCm38 | |
Downstream | predicted gene 16070 | Gm16070 | OTTMUSG00000029756 | 54.1kb | Sense | Chr. 1 - 17676027 - 17727646 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
