Assay Details
| Gene Symbol: Trmt61b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015549.1 | 3 - 4 | 1008 | 124 | ||
| NR_027952.1 | 3 - 4 | 1008 | 124 | |||
| GenBank mRNA | AK032413.1 | - | 3 - 4 | 993 | 124 | |
| AK036113.1 | - | 3 - 4 | 1002 | 124 | ||
| AK045842.1 | - | 3 - 4 | 954 | 124 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
methyltransferase like 4, pseudogene 1 |
Mettl4-ps1 | ENSMUSG00000079356 | -25.6kb | Anti-sense | Chr. 17 - 71528986 - 71544207 on NCBI Build GRCm38 |
| Downstream | WD repeat domain 43 | Wdr43 |
2610318G08Rik, AU020887, AV024208, Wrd43, mKIAA0007 |
26.9kb | Anti-sense | Chr. 17 - 71615900 - 71659031 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt61b
Genomic Map