Assay Details
| Gene Symbol: Slc3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009205.2 | NP_033231.2 | 2 - 3 | 691 | 146 | |
| GenBank mRNA | AK052623.1 | - | 2 - 3 | 662 | 146 | |
| AK132275.1 | - | 2 - 3 | 663 | 146 | ||
| BC013441.1 | - | 2 - 3 | 633 | 4222972 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc3a1
Genomic Map