Assay Details
Gene Symbol: Pfkfb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173019.5 | NP_766607.3 | 6 - 7 | 631 | 77 | |
GenBank mRNA | AK155172.1 | - | 6 - 7 | 569 | 77 | |
AY646092.2 | - | 6 - 7 | 517 | 77 | ||
AY707862.1 | - | 6 - 7 | 534 | 77 | ||
AY756065.1 | - | 6 - 7 | 517 | 77 | ||
AY756066.1 | - | 6 - 7 | 517 | 77 | ||
AY756067.1 | - | 6 - 7 | 517 | 77 | ||
BC057594.1 | - | 6 - 7 | 583 | 6810667 | 77 | |
CA316033.1 | - | 6 - 7 | 583 | 77 | ||
DQ350638.1 | - | 7 - 8 | 603 | 77 | ||
EU571215.1 | - | 6 - 7 | 631 | 77 | ||
EU571216.1 | - | 8 - 9 | 790 | 77 | ||
EU571217.1 | - | 6 - 7 | 758 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY756066.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
