Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900011788.mm.1 |
NM_173019.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900011788.mm.1 |
NM_173019.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900011788.mm.1 |
TC0900001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900011790.mm.1 |
NM_173019.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900011790.mm.1 |
NM_173019.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900011790.mm.1 |
TC0900001430.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900011789.mm.1 |
NM_173019.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900011789.mm.1 |
NM_173019.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900011789.mm.1 |
TC0900001430.mm.1 |
MTA TC |