Assay Details
| Gene Symbol: BC048562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004192.1 | NP_001004192.1 | 2 - 3 | 209 | 65 | |
| GenBank mRNA | AK029636.1 | - | 2 - 3 | 210 | 65 | |
| BC048562.1 | - | 2 - 3 | 209 | 6703719 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

BC048562
Genomic Map