Assay Details
| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011830.3 | NP_035960.2 | 6 - 7 | 652 | 71 | |
| GenBank mRNA | AK002676.1 | - | 6 - 7 | 684 | 71 | |
| AK146362.1 | - | 6 - 7 | 718 | 71 | ||
| AK150713.1 | - | 6 - 7 | 688 | 71 | ||
| AK151233.1 | - | 6 - 7 | 688 | 71 | ||
| AK151706.1 | - | 6 - 7 | 711 | 71 | ||
| AK152691.1 | - | 6 - 7 | 685 | 71 | ||
| AK159822.1 | - | 6 - 7 | 685 | 71 | ||
| AK160569.1 | - | 6 - 7 | 685 | 71 | ||
| AK166667.1 | - | 6 - 7 | 687 | 71 | ||
| AK169129.1 | - | 6 - 7 | 718 | 71 | ||
| AK169788.1 | - | 6 - 7 | 718 | 71 | ||
| BC010314.1 | - | 6 - 7 | 652 | 2646679 | 71 | |
| BC052671.1 | - | 6 - 7 | 673 | 30058399 | 71 | |
| M33934.1 | - | 6 - 7 | 685 | 71 | ||
| M98333.1 | - | 6 - 7 | 619 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Impdh2
Genomic Map