Assay Details
| Gene Symbol: Ndufaf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023247.1 | NP_075736.1 | 2 - 3 | 689 | 86 | |
| GenBank mRNA | AF235017.1 | - | 2 - 3 | 689 | 86 | |
| AK005252.1 | - | 2 - 3 | 304 | 86 | ||
| AK014634.1 | - | 2 - 3 | 304 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufaf3
Genomic Map