Assay Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281871.1 | NP_001268800.1 | 4 - 5 | 360 | 107 | |
| NM_026312.5 | NP_080588.1 | 4 - 5 | 360 | 107 | ||
| GenBank mRNA | AK007655.1 | - | 4 - 5 | 342 | 107 | |
| AK007872.1 | - | 3 - 4 | 229 | 107 | ||
| AK011610.1 | - | 4 - 5 | 324 | 107 | ||
| AK030507.1 | - | 4 - 5 | 342 | 107 | ||
| AK076889.1 | - | 4 - 5 | 342 | 107 | ||
| BC035042.1 | - | 4 - 5 | 324 | 1224959 | 107 | |
| BC044719.1 | - | 4 - 5 | 308 | 6394829 | 107 | |
| BC061070.1 | - | 4 - 5 | 321 | 6594922 | 107 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32579167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32579168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32579169 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | AK030507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | BC035042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | BC044719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | BC061070.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | NM_026312.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | AK030507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | BC035042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | BC044719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | BC061070.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P495171 | NM_026312.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213798 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29077 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144564 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | AK030507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | BC035042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | BC044719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | BC061070.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | NM_026312.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | AK030507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | BC035042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | BC044719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | BC061070.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P495171 | NM_026312.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32579180 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09383131 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09383160 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09383104 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09383117 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09383139 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144564 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09383178 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03731557 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03731541 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09383125 | - | - |
| Whole Mouse Genome G4122A | A_52_P272936 | AK030507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P272936 | NM_026312.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | AK030507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | BC035042.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | BC044719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | BC061070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | NM_026312.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | AK030507.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | BC035042.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | BC044719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | BC061070.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495171 | NM_026312.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P593861 | AK076889.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144564 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09383117 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14850209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14850204 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14850211 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14850203 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14850212 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14850213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32579167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32579168 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32579169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32579181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14743828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14513810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14542799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14676503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14513806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14513816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14556802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14850209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14676506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14850212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14636346 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14553018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14542798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15006605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32579178 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601339 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601337 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601336 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417921_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601335 | - | - |
| Mouse Genome 430 2.0 Array | 1417921_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417921_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67683_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67683_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00006982.mm.1 | NM_001281871.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006982.mm.1 | TC0X00001054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006981.mm.1 | NM_001281871.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006981.mm.1 | TC0X00001054.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00006973.mm.1 | NM_001281871.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006973.mm.1 | NM_026312.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00006973.mm.1 | TC0X00001054.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417921_PM_at | - | - |
| Mouse Expression Set 430 | 1417921_at | - | - |
| Murine Genome U74v2 Set | 161520_PM_at | - | - |
| Murine Genome U74v2 Set | 161520_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417921_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417921_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417921_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417921_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2650746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5420487 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420487 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605448 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694999 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694999 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2605448 | - | - |
| mouse-6 v1.1 (Illumina) | scl0067683.1_165-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002929.1_32-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650746 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420487 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002929.1_32-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067683.1_165-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650746 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420487 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2605448 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257485 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2834344 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2605448 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2834344 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257485 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0002929.1_32-S | - | - |

Pbdc1
Genomic Map