Assay Details
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144860.2 | NP_659109.2 | 2 - 3 | 1081 | 137 | |
GenBank mRNA | AK139338.1 | - | 2 - 3 | 962 | 137 | |
AY147848.1 | - | 2 - 3 | 580 | 137 | ||
AY149907.1 | - | 2 - 3 | 401 | 137 | ||
AY245539.1 | - | 2 - 3 | 1081 | 137 | ||
AY974091.1 | - | 2 - 3 | 374 | 137 | ||
BC131662.1 | - | 3 - 4 | 547 | 40110696 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
