Assay Details
| Gene Symbol: Mib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144860.2 | NP_659109.2 | 8 - 9 | 1917 | 85 | |
| GenBank mRNA | AK053035.1 | - | 1 - 2 | 140 | 85 | |
| AK080847.1 | - | 2 - 3 | 193 | 85 | ||
| AY147848.1 | - | 8 - 9 | 1416 | 85 | ||
| AY149907.1 | - | 8 - 9 | 1237 | 85 | ||
| AY245539.1 | - | 8 - 9 | 1917 | 85 | ||
| AY974091.1 | - | 8 - 9 | 1210 | 85 | ||
| BC131662.1 | - | 9 - 10 | 1383 | 40110696 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mib1
Genomic Map