Assay Details
| Gene Symbol: Gata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010258.3 | NP_034388.2 | 3 - 4 | 1557 | 73 | |
| GenBank mRNA | AF179425.1 | - | 2 - 3 | 864 | 73 | |
| AK142381.1 | - | 3 - 4 | 1417 | 73 | ||
| S82462.1 | - | 2 - 3 | 1319 | 73 | ||
| U51335.1 | - | 1 - 2 | 137 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gata6
Genomic Map