Assay Details
| Gene Symbol: Gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148331.1 | - | 2 - 3 | 293 | 164 | |
| AK193344.1 | - | 321 | 164 | |||
| BC047998.1 | - | 2 - 3 | 205 | 6511097 | 164 | |
| BC138310.1 | - | 2 - 3 | 186 | 8861330 | 164 | |
| BY741452.1 | - | 2 - 3 | 292 | 164 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148331.1, BC047998.1, BC138310.1 transcripts.
Set Membership: |
|

Gpatch11
Genomic Map