Assay Details
| Gene Symbol: Gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181649.6 | NP_857632.4 | 7 - 8 | 843 | 99 | |
| GenBank mRNA | AK009084.1 | - | 7 - 8 | 729 | 99 | |
| AK148331.1 | - | 7 - 8 | 845 | 99 | ||
| BC047998.1 | - | 7 - 8 | 757 | 6511097 | 99 | |
| BC092522.1 | - | 7 - 8 | 770 | 30648550 | 99 | |
| BC138310.1 | - | 7 - 8 | 738 | 8861330 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpatch11
Genomic Map