Assay Details
| Gene Symbol: Twf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011876.3 | NP_036006.1 | 1 - 2 | 177 | 81 | |
| GenBank mRNA | AK084026.1 | - | 1 - 2 | 177 | 81 | |
| AK155204.1 | - | 1 - 2 | 160 | 81 | ||
| AK157886.1 | - | 1 - 2 | 122 | 81 | ||
| AK166900.1 | - | 1 - 2 | 145 | 81 | ||
| AK170441.1 | - | 1 - 2 | 125 | 81 | ||
| BC003338.1 | - | 1 - 2 | 124 | 3257230 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084026.1, AK155204.1, AK157886.1, AK166900.1, AK170441.1, BC003338.1, NM_011876.3 transcripts.
Set Membership: |
|

Twf2
Genomic Map