Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024947.mm.1 |
NM_020559.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024947.mm.1 |
NM_020559.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024947.mm.1 |
TC0900003004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024949.mm.1 |
NM_020559.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024949.mm.1 |
NM_020559.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024949.mm.1 |
TC0900003004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024950.mm.1 |
NM_020559.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024950.mm.1 |
NM_020559.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024950.mm.1 |
TC0900003004.mm.1 |
MTA TC |