Assay Details
| Gene Symbol: Zbtb38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054529.1 | - | 1 - 2 | 85 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK054529.1 transcripts.
Set Membership: |
|

Zbtb38
Genomic Map