Assay Details
| Gene Symbol: Six3os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038084.1 | 2 - 3 | 488 | 98 | ||
| GenBank mRNA | AY590888.1 | - | 2 - 3 | 317 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Six3os1
Genomic Map