Assay Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011104.3 | NP_035234.1 | 12 - 13 | 2541 | 73 | |
GenBank mRNA | AF028009.1 | - | 12 - 13 | 1742 | 73 | |
AF325507.1 | - | 12 - 13 | 1733 | 73 | ||
AK082438.1 | - | 12 - 13 | 2542 | 73 | ||
AK085351.1 | - | 13 - 14 | 1953 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
