Assay Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011104.3 | NP_035234.1 | 4 - 5 | 1413 | 93 | |
GenBank mRNA | AF028009.1 | - | 4 - 5 | 614 | 93 | |
AF325507.1 | - | 4 - 5 | 605 | 93 | ||
AK082438.1 | - | 4 - 5 | 1414 | 93 | ||
AK085351.1 | - | 5 - 6 | 825 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
