Assay Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011104.3 | NP_035234.1 | 7 - 8 | 1777 | 95 | |
GenBank mRNA | AF028009.1 | - | 7 - 8 | 978 | 95 | |
AF325507.1 | - | 7 - 8 | 969 | 95 | ||
AK082438.1 | - | 7 - 8 | 1778 | 95 | ||
AK085351.1 | - | 8 - 9 | 1189 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
