Assay Details
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010137.3 | NP_034267.3 | 13 - 14 | 2605 | 85 | |
GenBank mRNA | AK136911.1 | - | 13 - 14 | 2609 | 85 | |
AK141185.1 | - | 13 - 14 | 2340 | 85 | ||
AK158430.1 | - | 13 - 14 | 2769 | 85 | ||
BC057870.1 | - | 13 - 14 | 2327 | 5032291 | 85 | |
D89787.1 | - | 13 - 14 | 2567 | 85 | ||
U81983.1 | - | 13 - 14 | 2288 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
