Assay Details
Gene Symbol: Epas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010137.3 | NP_034267.3 | 6 - 7 | 1192 | 63 | |
GenBank mRNA | AF045160.1 | - | 6 - 7 | 956 | 63 | |
AK141185.1 | - | 6 - 7 | 929 | 63 | ||
AK158430.1 | - | 6 - 7 | 1356 | 63 | ||
BC057870.1 | - | 6 - 7 | 914 | 5032291 | 63 | |
D89787.1 | - | 6 - 7 | 1154 | 63 | ||
U81983.1 | - | 6 - 7 | 875 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
